Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nalpha-(2,4-Dinitrophenyl)-L-arginine 98.0+%, TCI America™
SDP

Catalog No. D10401G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D10401G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D10401G Supplier TCI America Supplier No. D10401G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1602-42-2
Molecular Formula C12H16N6O6
Molecular Weight (g/mol) 340.30
MDL Number MFCD00038105
InChI Key GZJXZYUXRVBOAH-UHFFFAOYNA-N
Synonym Nalpha-Dnp-L-arginine, Dnp-Arg-OH
PubChem CID 7083742
IUPAC Name 5-[(diaminomethylidene)amino]-2-[(2,4-dinitrophenyl)amino]pentanoic acid
SMILES NC(N)=NCCCC(NC1=CC=C(C=C1[N+]([O-])=O)[N+]([O-])=O)C(O)=O

Specifications

Color Yellow
Quantity 1 g
UN Number 1325
Formula Weight 340.30
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Nalpha-(2,4-Dinitrophenyl)-L-arginine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.