Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nalpha-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-asparagine 98.0+%, TCI America™
SDP

Catalog No. F03065G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F03065G 5 g
F030625G 25 g
F03061G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. F03065G Supplier TCI America Supplier No. F03065G

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Chemical Identifiers

CAS 71989-16-7
Molecular Formula C19H18N2O5
Molecular Weight (g/mol) 354.362
MDL Number MFCD00037132
InChI Key YUGBZNJSGOBFOV-INIZCTEOSA-N
Synonym fmoc-asn-oh, fmoc-l-asn-oh, fmoc-l-asparagine, nalpha-fmoc-l-asparagine, n-fmoc-l-asparagine, fmoc-asn, nalpha-9h-fluoren-9-ylmethoxy carbonyl-l-asparagine, n-a-fmoc-l-asparagine, 9-fluorenylmethoxycarbonyl-l-asparagine, 2s-3-carbamoyl-2-9h-fluoren-9-ylmethoxy carbonyl amino propanoic acid
PubChem CID 2724774
IUPAC Name (2S)-4-amino-2-(9H-fluoren-9-ylmethoxycarbonylamino)-4-oxobutanoic acid
SMILES C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NC(CC(=O)N)C(=O)O

Specifications

Color White
Quantity 5 g
Formula Weight 354.36
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Nalpha-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-asparagine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.