Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nalpha-[(9H-Fluoren-9-ylmethoxy)carbonyl]-N1-tert-butoxycarbonyl-L-tryptophan 97.0+%, TCI America™
SDP

Catalog No. F050725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F050725G 25 g
F05075G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F050725G Supplier TCI America Supplier No. F050725G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 143824-78-6
Molecular Formula C31H30N2O6
Molecular Weight (g/mol) 526.59
MDL Number MFCD00153366
InChI Key ADOHASQZJSJZBT-SANMLTNESA-N
Synonym fmoc-trp boc-oh, fmoc-l-trp boc-oh, nalpha-9h-fluoren-9-ylmethoxy carbonyl-n1-tert-butoxycarbonyl-l-tryptophan, s-2-9h-fluoren-9-yl methoxy carbonyl amino-3-1-tert-butoxycarbonyl-1h-indol-3-yl propanoic acid, 2s-3-1-tert-butoxycarbonyl indol-3-yl-2-9h-fluoren-9-ylmethoxy carbonyl amino propanoic acid, fmoc-l-trp boc, pubchem10048, 1-boc-n-fmoc-l-tryptophan, ksc910o8t, n1-boc-nalpha-fmoc-l-tryptophan
PubChem CID 9849766
IUPAC Name (2S)-3-{1-[(tert-butoxy)carbonyl]-1H-indol-3-yl}-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)propanoic acid
SMILES CC(C)(C)OC(=O)N1C=C(C[C@H](NC(=O)OCC2C3=C(C=CC=C3)C3=C2C=CC=C3)C(O)=O)C2=CC=CC=C12

Specifications

Melting Point 99°C
Color White-Yellow
Quantity 25 g
Formula Weight 526.59
Percent Purity ≥97.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Nalpha-[(9H-Fluoren-9-ylmethoxy)carbonyl]-N1-tert-butoxycarbonyl-L-tryptophan
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.