Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nalpha-[(9H-Fluoren-9-ylmethoxy)carbonyl]-Nepsilon-(4-methyltrityl)-L-lysine 98.0+%, TCI America™
SDP

Catalog No. F10431G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F10431G 1 g
F10435G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F10431G Supplier TCI America Supplier No. F10431G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 167393-62-6
Molecular Formula C41H40N2O4
Molecular Weight (g/mol) 624.78
MDL Number MFCD00237166
InChI Key YPTNAIDIXCOZAJ-WHUAGQPENA-N
Synonym fmoc-lys mtt-oh, fmoc-l-lys mtt-oh, fmoc-n'-methyltrityl-l-lysine, nalpha-fmoc-nepsilon-4-methyltrityl-l-lysine, s-2-9h-fluoren-9-yl methoxy carbonyl amino-6-diphenyl p-tolyl methyl amino hexanoic acid, fmoc-lys meotrt-oh, fmoc-lysine mtt-oh, fmoc-l-lys mmt-oh, ambotzfaa1135, fmoc-lys ch3trt-oh
PubChem CID 10963195
IUPAC Name (2S)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-6-{[(4-methylphenyl)diphenylmethyl]amino}hexanoic acid
SMILES CC1=CC=C(C=C1)C(NCCCC[C@H](NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 140°C
Color White
Quantity 1 g
Formula Weight 624.78
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material Nalpha-[(9H-Fluoren-9-ylmethoxy)carbonyl]-Nepsilon-(4-methyltrityl)-L-lysine
TSCA No
Recommended Storage Freezer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.