Learn More
Nalpha-Benzyloxycarbonyl-L-ornithine, 98%
Description
Nalpha-Benzyloxycarbonyl-L-ornithine is widely used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Nalpha-Benzyloxycarbonyl-L-ornithine is widely used as pharmaceutical intermediate.
Solubility
Soluble in 0.1N NaOG (1 mg/ml-clear, colorless solution). Slightly soluble in water.
Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
Specifications
Specifications
| Color | White |
| Solubility Information | Soluble in 0.1N NaOG (1mg/ml-clear,colorless solution). Slightly soluble in water. |
| Quantity | 1 g |
| Formula Weight | 266.29 |
| Percent Purity | 98% |
| Physical Form | Powder |
| Chemical Name or Material | Nalpha-Benzyloxycarbonyl-L-ornithine |
Safety and Handling
| TSCA | No |
| storageNote1 | 2° to 8°C |
| Recommended Storage | Keep cold |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.