Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nalpha-Carbobenzoxy-D-tryptophan 98.0+%, TCI America™
SDP

Catalog No. C21301G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C21301G 1 g
C21305G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C21301G Supplier TCI America Supplier No. C21301G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2279-15-4
Molecular Formula C19H18N2O4
Molecular Weight (g/mol) 338.36
MDL Number MFCD00037945
InChI Key AHYFYYVVAXRMKB-QGZVFWFLSA-N
Synonym z-d-trp-oh, n-cbz-d-tryptophan, z-d-tryptophan, cbz-d-trp-oh, nalpha-cbz-d-tryptophan, r-2-benzyloxy carbonyl amino-3-1h-indol-3-yl propanoic acid, n a-benzyloxycarbonyl-d-tryptophan, chembl65134, nalpha-carbobenzoxy-d-tryptophan, d-tryptophan, n-phenylmethoxy carbonyl
PubChem CID 700663
IUPAC Name (2R)-2-{[(benzyloxy)carbonyl]amino}-3-(1H-indol-3-yl)propanoic acid
SMILES OC(=O)[C@@H](CC1=CNC2=CC=CC=C12)NC(=O)OCC1=CC=CC=C1

Specifications

Melting Point 123°C
Color White-Yellow
Quantity 1 g
Formula Weight 338.36
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Nalpha-Carbobenzoxy-D-tryptophan
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.