Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nalpha-Carbobenzoxy-L-histidine 99.0+%, TCI America™
SDP

Catalog No. B026525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B026525G 25 g
B02651G 1 g
B02655G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B026525G Supplier TCI America Supplier No. B026525G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 14997-58-1
Molecular Formula C14H15N3O4
Molecular Weight (g/mol) 289.291
MDL Number MFCD00065960
InChI Key WCOJOHPAKJFUDF-LBPRGKRZSA-N
Synonym z-his-oh, n-cbz-l-histidine, cbz-his-oh, cbz-l-his-oh, nalpha-cbz-l-histidine, cbz-histidine, nalpha-carbobenzoxy-l-histidine, l-histidine, n-phenylmethoxy carbonyl, z-l-histidine, s-2-benzyloxy carbonyl amino-3-1h-imidazol-4-yl propanoic acid
PubChem CID 736313
IUPAC Name (2S)-3-(1H-imidazol-5-yl)-2-(phenylmethoxycarbonylamino)propanoic acid
SMILES C1=CC=C(C=C1)COC(=O)NC(CC2=CN=CN2)C(=O)O

Specifications

Melting Point 163°C
Color White
Quantity 25 g
Formula Weight 289.29
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Nalpha-Carbobenzoxy-L-histidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.