Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nalpha-Fmoc-Nepsilon-(4-methyltrityl)-L-lysine, 95%

Catalog No. aah6362903
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6362903 1 g
AAH6362906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6362903 Supplier Thermo Scientific Chemicals Supplier No. H6362903
Only null left
Add to Cart

CAS: 167393-62-6 | C41H40N2O4 | 624.78 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 167393-62-6
Molecular Formula C41H40N2O4
Molecular Weight (g/mol) 624.78
MDL Number MFCD00237166
InChI Key YPTNAIDIXCOZAJ-WHUAGQPENA-N
Synonym fmoc-lys mtt-oh, fmoc-l-lys mtt-oh, fmoc-n'-methyltrityl-l-lysine, nalpha-fmoc-nepsilon-4-methyltrityl-l-lysine, s-2-9h-fluoren-9-yl methoxy carbonyl amino-6-diphenyl p-tolyl methyl amino hexanoic acid, fmoc-lys meotrt-oh, fmoc-lysine mtt-oh, fmoc-l-lys mmt-oh, ambotzfaa1135, fmoc-lys ch3trt-oh
PubChem CID 10963195
IUPAC Name (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-6-[[(4-methylphenyl)-diphenylmethyl]amino]hexanoic acid
SMILES CC1=CC=C(C=C1)C(NCCCC[C@H](NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Quantity 1 g
Formula Weight 624.77
Percent Purity 95%
Physical Form Solid
Chemical Name or Material Nalpha-Fmoc-Nepsilon-(4-methyltrityl)-L-lysine
TSCA No
storageNote1 -30° to -10°C
Recommended Storage Store at -20°C

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.