Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nalpha-Fmoc-Nepsilon-trifluoracetyl-L-lysine, 95%, Thermo Scientific™

Catalog No. AAH6224303
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6224303 1 g
AAH6224306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6224303 Supplier Thermo Scientific Chemicals Supplier No. H6224303

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 76265-69-5
Molecular Formula C23H23F3N2O5
Molecular Weight (g/mol) 464.44
MDL Number MFCD00153360
InChI Key ZVLMWTPNDXNXSZ-TWYLJJHKNA-N
Synonym fmoc-lys tfa-oh, fmoc-n-epsilon-trifluoroacetyl-l-lysine, n-alpha-9-fluorenylmethyloxycarbonyl-n-epsilon-trifluoracetyl-l-lysine, fmoc-lys-tfa-oh, n, a-fmoc-n, a-trifluoroacetyl-l-lysine, fmoc-lys tfa-oh hplc, 2s-2-9h-fluoren-9-ylmethoxycarbonylamino-6-2,2,2-trifluoroacetyl amino hexanoic acid, n∼2∼-9h-fluoren-9-yl methoxy carbonyl-n∼6∼-trifluoroacetyl-l-lysine, n2-9h-fluoren-9-yl methoxy carbonyl-n6-2,2,2-trifluoroacetyl-l-lysine, 2s-2-9h-fluoren-9-ylmethoxy carbonyl amino-6-2,2,2-trifluoroacetamido hexanoic acid
PubChem CID 16213176
IUPAC Name (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-6-[(2,2,2-trifluoroacetyl)amino]hexanoic acid
SMILES OC(=O)[C@H](CCCCNC(=O)C(F)(F)F)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12

Specifications

Melting Point 170°C to 174°C
Quantity 1 g
Formula Weight 464.43
Percent Purity 95%
Physical Form Solid
Chemical Name or Material Nalpha-Fmoc-Nepsilon-trifluoracetyl-L-lysine
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.