Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nalpha-(tert-Butoxycarbonyl)-L-histidine 98.0+%, TCI America™
SDP

Catalog No. B163425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B163425G 25 g
B16345G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B163425G Supplier TCI America Supplier No. B163425G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 17791-52-5
Molecular Formula C11H17N3O4
Molecular Weight (g/mol) 255.274
MDL Number MFCD00065576
InChI Key AYMLQYFMYHISQO-QMMMGPOBSA-N
Synonym boc-his-oh, n-boc-l-histidine, boc-l-histidine, tert-butyloxycarbonyl-l-histidine, nalpha-tert-butoxycarbonyl-l-histidine, nalpha-boc-l-histidine, boc-l-his-oh, n-tert-butoxycarbonyl-l-histidine, l-histidine, n-1,1-dimethylethoxy carbonyl
PubChem CID 87308
IUPAC Name (2S)-3-(1H-imidazol-5-yl)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
SMILES CC(C)(C)OC(=O)NC(CC1=CN=CN1)C(=O)O

Specifications

Color White
Quantity 25 g
Formula Weight 255.27
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Nalpha-(tert-Butoxycarbonyl)-L-histidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.