Learn More
AdipoGen anti-Nampt (Visfatin/PBEF), mAb (OMNI379) (100 µg)
SDP
Supplier:  AdipoGen AG20A0034C100
Encompass_Preferred
Monoclonal Antibody. Recognizes human, mouse and rat Nampt. Isotype: Mouse IgG2akappa. Clone: OMNI379. Applications: ELISA, FACS, ICC, IHC, IP, WB. Liquid. 0.2µm-filtered solution in PBS, pH 7.4. Contains no preservatives. Nicotinamide phosphoribosyltransferase (Nampt; pre-B cell colony-enhancing factor; PBEF; Visfatin) is an 52kDa adipokine secreted by adipose tissue and involved in the biosynthesis of nicotinamide adenine dinucleotide (NAD+). Two forms of Nampt exist, an intracellular form (iNampt) and an extracellular form (eNampt). While the function of iNampt as an essential and rate-limiting NAD+ biosynthetic enzyme is well established, the physiological role of eNampt is still a matter of debate. Nampt has various functions, including the promotion of vascular smooth muscle cell maturation and inhibition of neutrophil apoptosis. It activates insulin receptor and has insulin-mimetic effects, lowering blood glucose and improving insulin sensitivity.
Catalog No. 50-205-2822
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.