Learn More
Medchemexpress LLC Nanatinostat | 1256448-47-1 | 99.7% | 394.40 | C20H19FN6O2 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY134325MG
Encompass_Preferred
Nanatinostat is a second-generation hydroxamic acid-based inhibitor of class I histone deacetylases (HDAC1/2/3). It is orally bioavailable, shows low-nanomolar potency and selectivity, induces apoptosis in cancer cells, and is supplied for research use as a high-purity solid or as a DMSO solution.
  • Provides potent inhibition of HDAC1, HDAC2, and HDAC3 at low-nanomolar concentrations.
  • Exhibits reduced activity against HDAC5 and HDAC6, improving selectivity.
  • Induces apoptosis and antiproliferative effects in cancer cell models.
  • Suitable for in vitro and in vivo epigenetic and anticancer studies.
  • Available as a high-purity solid and as a DMSO solution for flexible dosing.
  • High purity supports reproducible biochemical and cell-based assays.
Catalog No. 50-002-22810