Learn More
Medchemexpress LLC Nanatinostat | 1256448-47-1 | 99.7% | C20H19FN6O2 | 10MG
SDP
Supplier:  Medchemexpress LLC HY1343210MG
Encompass_Preferred
Nanatinostat is an orally bioavailable hydroxamic acid-based histone deacetylase (HDAC) inhibitor with reported antiproliferative and pro-apoptotic activity in cancer cell lines. Supplied as a research-grade compound with high purity, it is used in oncology and epigenetics studies and is available as solid vials and DMSO solution formats.
  • Inhibits histone deacetylase (HDAC) activity.
  • Demonstrates antiproliferative activity in cancer cell lines.
  • Shows pro-apoptotic effects and induces cell cycle arrest.
  • High purity suitable for research applications.
  • Available as solid vials and DMSO solution for flexible dosing.
Catalog No. 50-002-61747