Learn More
TARGETMOL CHEMICALS INC Nandrolone decanoate 200MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T2556200MG
Encompass_Preferred
Also available in 1 mL, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. Nandrolone decanoate (19-Nortestosterone decanoate) is the decanoate salt form of nandrolone, an anabolic steroid analog of testosterone with androgenic, anabolic, and erythropoietin-stimulating effects. Nandrolone binds to and activates specific nuclear androgen receptors in responsive tissues, including the prostate, seminal vesicles, scrotum, penis, larynx, hair follicles, muscle, and bone. Purity 99.68%
Catalog No. 50-287-9191
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.