Learn More
Promega Nano-Glo™ Dual-Luciferase™ Reporter Assay/pNL1.1.TK Bundle
Catalog No. PRN1521 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
1 Ea.
Catalog No. Quantity
PRN1521 1 Ea.
1 options
Catalog No. PRN1521 Supplier Promega Supplier No. N1521
The Nano-Glo™ Dual-Luciferase Reporter (NanoDLR) Assay System is a homogeneous reagent system that allows you to sequentially detect the activities of firefly (Photinus pyralis) luciferase and NanoLuc luciferase (Nluc) from a single sample.
  • Ultra-Sensitive Detection of Firefly and NanoLuc(R) Luciferase Activities in a Single Sample
  • Greater sensitivity at low expression levels
  • Choose either firefly or NanoLuc(R) luciferase as primary reporters
  • Improved reagent stability brings more flexibility to assay design
Specifications
Quantity 1 Ea.
Content And Storage Varies Among Components