Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega Nano-Glo™ Dual-Luciferase™ Reporter Assay System

Catalog No. PRN1650 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
10 mL
100 mL
10 x 10 mL
10 x 100 mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRN1650 10 x 100 mL
PRN1610 10 mL
PRN1620 100 mL
PRN1630 10 x 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. PRN1650 Supplier Promega Supplier No. N1650
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

The Nano-Glo™ Dual-Luciferase Reporter (NanoDLR) Assay System is a homogeneous reagent system that allows you to sequentially detect the activities of firefly (Photinus pyralis) luciferase and NanoLuc luciferase (Nluc) from a single sample.

  • Ultra-Sensitive Detection of Firefly and NanoLuc(R) Luciferase Activities in a Single Sample
  • Greater sensitivity at low expression levels
  • Choose either firefly or NanoLuc(R) luciferase as primary reporters
  • Improved reagent stability brings more flexibility to assay design

Specifications

Quantity 10 x 100 mL
Content And Storage -30°C to -10°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.