Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega Nano-Glo HiBiT Blotting System, 100ml

Catalog No. prn2410 Shop All Promega Corporation Products
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRN2410 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PRN2410 Supplier Promega Supplier No. N2410
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Bioluminescent detection of HiBiT tagged proteins on nitrocellulose membranes without antibodies.

  • Fast Luminescent Detection of Proteins on Blots
  • Determine protein size and quantify expression on blots
  • Protocol requires only minutes, with few processing steps
  • Femtogram sensitivity proportional over five orders of magnitude

Specifications

Content And Storage -30°C to -10°C
Quantity 100 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.