Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega Nano-Glo HiBiT Extracellular Detection System, 100ml

Catalog No. PRN2421 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Nano-Glo HiBiT Extracellular Detection System, 100ml
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
PRN2421 Nano-Glo HiBiT Extracellular Detection System, 100ml
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PRN2421 Supplier Promega Supplier No. N2421
May include imposed supplier surcharges.
Add to Cart
Add to Cart
This item is not returnable. View return policy

Nano-Glo HiBiT Extracellular Detection System quantitates cell surface or secreted protein expression using a simple add-mix-read assay format and the nonlytic Nano-Glo HiBiT Extracellular Detection Reagent.

Simple Bioluminescent Detection of Cell-Surface Protein Expression.
The Nano-Glo HiBiT Extracellular Detection System quantitates cell surface or secreted protein expression in minutes using a simple add-mix-read assay format. Using the nonlytic Nano-Glo™ HiBiT Extracellular Detection Reagent, any proteins that are tagged with the 11 amino acid HiBiT peptide and expressed outside of the cell can be specifically quantitated. The detection reagent contains the complementary polypeptide LgBiT, which spontaneously interacts with the HiBiT tag to reconstitute the bright, luminescent NanoBiT™ enzyme. Luminescence is proportional to the amount of HiBiT-tagged protein present outside of the cell over seven orders of magnitude.

  • Quantify HiBiT-Tagged Proteins Expressed on the Cell Surface
  • Specific, live-cell detection of extracellular expressed or secreted proteins
  • Simple add-mix-read assay format—no antibodies required
  • Quantitate over 7 logs of linear dynamic range
  • Each system contains sufficient components to prepare 100ml reagent

Includes:

  • Nano-Glo HiBiT Extracellular Buffer, N243B (100mL)
  • Nano-Glo HiBiT Extracellular Substrate, N300B (2 x 1mL)
  • LgBiT Protein, N401B (1mL)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.