Learn More
Promega Nano-Glo HiBiT Extracellular Detection System, 10ml
Catalog No. PRN2420 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Nano-Glo HiBiT Extracellular Detection System, 10ml
Catalog No.
PRN2420 Nano-Glo HiBiT Extracellular Detection System, 10ml
1 options
This item is not returnable. View return policy
Catalog No. PRN2420 Supplier Promega Supplier No. N2420
This item is not returnable. View return policy
Nano-Glo HiBiT Extracellular Detection System quantitates cell surface or secreted protein expression using a simple add-mix-read assay format and the nonlytic Nano-Glo HiBiT Extracellular Detection Reagent.
  • Quantify HiBiT−Tagged Proteins Expressed on the Cell Surface
  • Specific, live−cell detection of extracellular expressed or secreted proteins
  • Simple add−mix−read assay format—no antibodies required
  • Quantitate over 7 logs of linear dynamic range