Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega Nano-Glo HiBiT Lytic Detection System, 10 x 100ml

Catalog No. PRN3050 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
10 x 100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRN3050 10 x 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PRN3050 Supplier Promega Supplier No. N3050
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

Accurately quantify Any HiBiT Tagged Protein Expressed in Cells in an antibody free, plate based method.

  • Accurately Quantify Any HiBiT−Tagged Protein Expressed in Cells
  • Sensitive bioluminescent protein detection
  • Simple add−and−read assay—no antibodies required
  • Quantitate over 7 logs of linear dynamic range

Specifications

Quantity 10 x 100 mL
Content And Storage -30°C to -10°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.