Learn More
Proteintech Group Inc Nano-Secondary alpaca anti-human IgG/anti-rabbit IgG, recombinant VHH, for 2x Cys conjugation [CTK0101]
SDP
Supplier:  Proteintech Group Inc SRBGCYS21500
Encompass
Monovalent, recombinant secondary single domain antibody to human IgG and rabbit IgG: alpaca monoclonal Nanobody, Fc- specific, for cysteine conjugation with thiol-reactive reagents, e.g. maleimides
Catalog No. 50-225-8860
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.