Learn More
CTL Scientific Supply Corp NANOCOLOR Zirconium 100 , contains 20 tube tests
SDP
Supplier:  CTL Scientific Supply Corp 985001
Encompass_Preferred
Tube test for the determination of Zirconium. Precise rapid tests for all kind of water and waste water samples. Time-saving and reliable analysis together with Macherey-Nagel NANOCOLOR photometers. Measuring range: Zirconium - 5–100 mg/L ZrManufactured by Macherey-Nagel
Catalog No. NC2958222