Learn More
Promega NanoBRET™ BRD4/Histone H3.3 Interaction Assay
Catalog No. PRN1830 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
1/Ea.
Catalog No. Quantity
PRN1830 1/Ea.
1 options
Catalog No. PRN1830 Supplier Promega Supplier No. N1830
Add to Cart
Add to Cart
Request a Quote
The NanoBRET Bromodomain BRD4/Histone Interaction Assays enable interaction studies of BRD-containing proteins with full-length histones in the context of natural chromatin.
  • Protein Interaction Assays to Study BRD/Histone Interactions in Live Cells
  • Study full-length bromodomain/histone-containing proteins or fragments
  • Detect protein interactions in live cells, even at low expression levels
  • Choose from prebuilt or custom assay options
Specifications
Content And Storage -30°C to -10°C
Quantity 1/Ea.