Learn More
Promega NanoBRET™ cMyc/MAX Interaction Assay
Catalog No. PRN1870 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
1/Ea.
Catalog No. Quantity
PRN1870 1/Ea.
1 options
Catalog No. PRN1870 Supplier Promega Supplier No. N1870
Add to Cart
Add to Cart
Request a Quote
NanoBRET Transcriptional Protein Assays are sensitive, reproducible live-cell assays designed for monitoring or screening the interaction of proteins involved in transcriptional regulation.
  • Sensitive Live-Cell Assays for Monitoring Proteins Interactions in Transcription Regulation
  • Order NanoBRET™ cMyc/MAX Interaction Assay by selecting N1870
  • For additional transcriptional assays go to View Prebuilt Assays link below
Specifications
Content And Storage -30°C to -10°C
Quantity 1/Ea.