Learn More
Promega NanoBRET™ Nano-Glo™ Detection System
Catalog No. p-7068002 Shop All Promega Corporation Products
Change view
Click to view available options
Quantity:
10,000 Assays
1000 Assays
200 Assays
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRN1661 200 Assays
PRN1662 1000 Assays
PRN1663 10,000 Assays
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. PRN1661 Supplier Promega Supplier No. N1661
Only null left
Add to Cart
Add to Cart
The NanoBRET Nano-Glo™ Detection System provides the NanoBRET Nano-Glo™ Substrate used by NanoLuc Luciferase to generate the donor signal and the HaloTag™ NanoBRET 618 Ligand for the fluorescent energy acceptor.
  • Dedicated Detection for NanoBRET™ Protein Interaction Assays
  • Measure protein interactions in live cells with full-length proteins
  • Study both on and off protein interactions with a reversible assay
  • Increased signal and lower background compared to conventional BRET assays
Specifications
Content And Storage -30°C to -10°C
Quantity 200 Assays