Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega NanoBRET™ Nano-Glo™ Substrate

Catalog No. prn1573 Shop All Promega Corporation Products
Change view
Click to view available options
Quantity:
50 μL
5 x 50 μL
2 x 1.25 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRN1573 2 x 1.25 mL
PRN1572 5 x 50 μL
PRN1571 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. PRN1573 Supplier Promega Supplier No. N1573
Only null left
Add to Cart
Add to Cart
Request a Quote

The NanoBRET™ Nano-Glo(R) Substrate is the substrate used by NanoLuc(R) Luciferase to generate the donor signal in several NanoBRET™ Assays.

For use with NanoBRET assay, a proximity-based assay that can detect protein interactions by measuring energy transfer from a bioluminescent protein donor to a fluorescent protein acceptor.

Specifications

Content And Storage -30°C to -10°C
Quantity 2 x 1.25 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.