Learn More
Promega NanoBRET™ Positive Control
Catalog No. PRN1581 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
2 x 20 μg
Catalog No. Quantity
PRN1581 2 x 20 μg
1 options
Catalog No. PRN1581 Supplier Promega Supplier No. N1581
The NanoBRET Positive Control Vector encodes a NanoLuc-HaloTag™ fusion protein that tethers together the NanoLuc donor and HaloTag™ acceptor proteins to ensure efficient energy transfer.
  • NanoLuc™-HaloTag™ fusion gene
  • Kanamycin resistance gene for selection in bacteria
  • A CMV promoter for high-level expression of NanoLuc™-HaloTag™ fusion in mammalian cells
The NanoBRET™ Positive Control Vector should be diluted in Transfection Carrier DNA to reduce cellular expression levels due to the high NanoBRET™ signal produced. The control vector can be used when performing instrument setup or as a reference control when performing other NanoBRET™ assays.
Specifications
Content And Storage -30°C to -10°C
Quantity 2 x 20 μg