Learn More
Promega NanoBRET™ PPI Flexi™ Starter System
Catalog No. PRN1821 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
1/Ea.
Catalog No. Quantity
PRN1821 1/Ea.
1 options
Catalog No. PRN1821 Supplier Promega Supplier No. N1821
Protein:Protein Interaction (PPI) Assays use NanoLuc Luciferase and HaloTag™ protein fused to target proteins of interest to enable sensitive, reproducible detection of protein interactions in the natural cellular environment.
  • BRET-Based Assays to Detect Protein:Protein Interactions in Live Cells
  • Study induction and inhibition of protein interactions using full-length proteins expressed at physiologically relevant levels
  • Starter Systems provide fusion vectors, positive controls and detection reagents for building PPI assays
  • Choice of multicloning site (MCS) or Flexi(R) vector-based cloning options
Specifications
Content And Storage COMPONENTS AT DIFFERENT TEMPS
Quantity 1/Ea.