Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega NanoBRET TE HDAC DNA Bundle, 1 each

Catalog No. PRN2120 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
1 Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRN2120 1 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PRN2120 Supplier Promega Supplier No. N2120
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

NanoBRET Target Engagement Assay Components are for use with all NanoBRET Target Engagement Assays.

  • For Use With All NanoBRET™ TE Assays
  • DNA Bundles and Transfection Carrier DNA
  • Intracellular TE Nano−Glo(R) Substrate/Inhibitor
  • Tracer Dilution Buffer

Specifications

Content And Storage -30°C to -10°C
Quantity 1 Each
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.