Learn More
Promega NanoBRET TE Intracellular Kinase Assay, K-4, 100 assays
Catalog No. PRN2520 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
100 Assays
Catalog No. Quantity
PRN2520 100 Assays
1 options
Catalog No. PRN2520 Supplier Promega Supplier No. N2520
Only null left
Add to Cart
Add to Cart
The NanoBRET Target Engagement (TE) Assay measures compound binding at select kinase targets within intact cells. The assay is based on the NanoBRET System, an energy transfer technique designed to measure molecular proximity in living cells.
  • Measure Compound Binding at Kinase Targets in Intact Cells
  • Detect kinase target engagement in live cells
  • Choose from >125 kinase fusion vectors
  • See Kinase Table (below) for available kinases and related information
Specifications
Quantity 100 Assays
Storage Requirements <−65°C