Learn More
Promega NanoBRET TE Intracellular Kinase Assay, K-5, 1,000 assays
Catalog No. PRN2501 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
1000 Assays
Catalog No. Quantity
PRN2501 1000 Assays
1 options
This item is not returnable. View return policy
Catalog No. PRN2501 Supplier Promega Supplier No. N2501
Add to Cart
Add to Cart
Request a Quote
This item is not returnable. View return policy
The NanoBRET Target Engagement (TE) Assay measures compound binding at select kinase targets within intact cells. The assay is based on the NanoBRET System, an energy transfer technique designed to measure molecular proximity in living cells.
  • Measure Compound Binding at Kinase Targets in Intact Cells
  • Detect kinase target engagement in live cells
  • Choose from >125 kinase fusion vectors
  • See Kinase Table (below) for available kinases and related information
Specifications
Quantity 1000 Assays
Storage Requirements <−65°C