Learn More
Therapak™ NanoCool™ Refrigerated Specimen Shipper
Catalog No. 22130418 Shop All Therapak Corporation Products
Change view
Click to view available options
Includes:
Sturdy transport box
Transport box
Quantity:
Up to 18 standard 13 x 100mm tubes
Up to 36 standard 13 x 100mm tubes
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes Quantity
22-130-418 Sturdy transport box Up to 36 standard 13 x 100mm tubes
22-130-417 Transport box Up to 18 standard 13 x 100mm tubes
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 22-130-418 Supplier Therapak™ Supplier No. 56850
Add to Cart
Add to Cart
This item is not returnable. View return policy
Includes
Sturdy transport box.
No gel refrigerants required
  • Portable unit maintains temperature from 2-8°C — ideal for overnight or international shipments
  • Weighs less and requires less space than conventional polystyrene boxes and ice packs, reducing shipping costs
  • Vacuum Insulated Panel (VIP) compartment transfers heat by evaporating small quantities of water at low pressure; a heat absorption method that's seven times more efficient than ice
  • Separate NanoCool cooler fits atop the VIP compartment and can be activated at room temperature with the push of a button
  • Small shipper holds up to 18 standard (13 × 100mm) transport tubes; large shipper holds up to 36 tubes
Order Info
One-time use only; cannot be reconditioned
Specifications
Quantity Up to 36 standard 13 x 100mm tubes
Includes Sturdy transport box
Type Portable Unit
Duration For overnight or international shipping