Learn More
World Precision Instrument Nanofil Application Kits-RPE-KIT
SDP
Catalog No. NC1485368
Encompass
These kits are specifically designed for ocular research applications, such as retinal pigment epithelium (RPE) and intraocular (IO) injection. The kits integrate into our NanoFil™ syringe, and can be paired with our UMP3 microinjection pump for accurate and repeatable submicroliter sample delivery. 
Catalog No. NC1485368 Supplier World Precision Instrument Supplier No. RPEKIT
These kits are specifically designed for ocular research applications, such as retinal pigment epithelium (RPE) and intraocular (IO) injection. The kits integrate into our NanoFil™ syringe, and can be paired with our UMP3 microinjection pump for accurate and repeatable submicroliter sample delivery. 