Learn More
R&D Systems Assay Plate/Lid Kit for Peggy/Sally, Peggy Sue/Sally Sue, or NanoPro 1000, 5/pk
SDP
Catalog No. NC9039079 Shop All Novus Biologicals Products
Encompass
Includes five 384-well plates and lids. Must be ordered with p/n 041-528.
Catalog No. NC9039079 Supplier R&D Systems Supplier No. 040663
Add to Cart
Add to Cart
Includes five 384-well plates and lids. Must be ordered with p/n 041-528.