Learn More
Sartorius Nanowell S200-100 100k
Catalog No. 502891582 Shop All Sartorius Products
Encompass
CellCelector Nanowell Plate 4 nl 24 wells
Catalog No. 50-289-1582 Supplier Sartorius Supplier No. CC0079
CellCelector Nanowell Plate 4 nl 24 wells