Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Naphthalene

Catalog No. 501858218
Encompass
Change view
Click to view available options
Quantity:
5 g
250 g
1 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-185-8218 250 g
50-185-8217 1 kg
50-185-8219 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 50-185-8218 Supplier Sigma Aldrich Supplier No. 184500250G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Naphthalene
TRUSTED_SUSTAINABILITY

Specifications

CAS 91-20-3
Melting Point 80°C to 82°C (lit.)
Boiling Point 218°C (lit.)
Molecular Formula C10H8
Linear Formula C10H8
MDL Number MFCD00001742
Quantity 250 g
Molecular Weight (g/mol) 128.17
Percent Purity ≥99%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.