Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Naphthol Yellow S Hydrate 98.0+%, TCI America™
SDP

Catalog No. F010025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F010025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F010025G Supplier TCI America Supplier No. F010025G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Laboratory reagent used in analytical applications.

  • Solubility: Soluble in water; slightly soluble in ethanol
  • Hazard Classification: Skin sensitization, Category 1 (H317)

Chemical Identifiers

CAS 846-70-8
Molecular Formula C10H4N2Na2O8S
Molecular Weight (g/mol) 358.188
MDL Number MFCD00003958
InChI Key CTIQLGJVGNGFEW-UHFFFAOYSA-L
Synonym 2,4-Dinitro-1-naphthol-7-sulfonic Acid Disodium Salt, Disodium 2,4-Dinitro-1-naphthol-7-sulfonate, Disodium 8-Hydroxy-5,7-dinitro-2-naphthalenesulfonate, 8-Hydroxy-5,7-dinitro-2-naphthalenesulfonic Acid Disodium Salt, Flavianic Acid Disodium Salt, Acid Ye
PubChem CID 2724063
ChEBI CHEBI:87219
IUPAC Name disodium;5,7-dinitro-8-oxidonaphthalene-2-sulfonate
SMILES C1=CC2=C(C=C1S(=O)(=O)[O-])C(=C(C=C2[N+](=O)[O-])[N+](=O)[O-])[O-].[Na+].[Na+]

Specifications

Color Pale Yellow to Yellow Red
Quantity 25 g
Physical Form Crystal/Powder
Formula Weight 358.19
Percent Purity ≥98.0% (HPLC,T)
Grade Laboratory
Chemical Name or Material Naphthol Yellow S Hydrate
EINECSNumber (5)-1392
RTECSNumber QK1813000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.