Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Naphthol Yellow S, Thermo Scientific™

Catalog No. AAB2087218
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2087218 50 g
AAB2087230 250 g
AAB208720B 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2087218 Supplier Thermo Scientific Chemicals Supplier No. B2087218

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 846-70-8
Molecular Formula C10H4N2Na2O8S
Molecular Weight (g/mol) 358.188
MDL Number MFCD00003958
InChI Key CTIQLGJVGNGFEW-UHFFFAOYSA-L
Synonym C.I. 10316; 5,7-Dinitro-8-hydroxy-2-naphthalenesulfonic acid disodium salt
PubChem CID 2724063
ChEBI CHEBI:87219
IUPAC Name disodium;5,7-dinitro-8-oxidonaphthalene-2-sulfonate
SMILES C1=CC2=C(C=C1S(=O)(=O)[O-])C(=C(C=C2[N+](=O)[O-])[N+](=O)[O-])[O-].[Na+].[Na+]

Specifications

Melting Point >300°C
Beilstein 2708476
Merck Index 14,6393
Formula Weight 358.2
Quantity 50 g
Chemical Name or Material Naphthol Yellow S
EINECSNumber 212-690-2
RTECSNumber QK1813000
TSCA Yes
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.