Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Naphtholbenzein Solution R, EP Reagent 1057601, Ricca Chemical
SDP

Catalog No. R5227500500 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R5227500500 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R5227500500 Supplier Ricca Chemical Company Supplier No. R5227500500C
Add to Cart
Add to Cart

Chemical Identifiers

CAS 64-19-7 , 145-50-6
Molecular Formula C2H4O2
Molecular Weight (g/mol) 60.05
MDL Number MFCD00036152
InChI Key QTBSBXVTEAMEQO-UHFFFAOYSA-N
Synonym ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure
PubChem CID 176
ChEBI CHEBI:15366
IUPAC Name acetic acid
SMILES CC(O)=O

Specifications

Color Red
CAS Min % 98.0, 0.19
CAS Max % 100.0, 0.19
Grade Laboratory
Packaging Amber Glass Bottle
pH <1
Quantity 500 mL
Physical Form Liquid
Chemical Name or Material Naphtholbenzein Solution, Reagent
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.