Learn More
Naphthylethylenediamine Dihydrochloride, 0.1% (w/v), Ricca Chemical
Shop All Ricca Chemical Company Products

Specifications
Specifications
| CAS | 7732-18-5 |
| Molecular Weight (g/mol) | 259.17 g/mol |
| Grade | Laboratory |
| Packaging | Amber Glass Bottle |
| Quantity | 500 mL |
| Chemical Name or Material | Naphthylethylenediamine Dihydrochloride Solution |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.