Learn More
Cayman Chemical Naproxen 5g
SDP
Supplier:  Cayman Chemical 70290 5G
Encompass_Preferred
A BCRP inhibitor sensitizes mitoxantrone-resistant H460/MX20 cells to mitoxantrone-induced cell death alone and in combination with UV radiation sensitizes KB-C2 cells overexpressing P-gp to colchicine-induced cell death increases intracellular accumulation of mitoxantrone in H460/MX20 cells at 5 UM
Catalog No. 50-289-6538
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.