Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medique Naproxen Sodium Tablets

Catalog No. 19090787 Shop All Medique Products
Change view
Click to view available options
Quantity:
50 x 1/Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
19-090-787 50 x 1/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-090-787 Supplier Medique Supplier No. 23750
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For long-lasting relief from pain associated with headaches, muscular aches and minor arthritis.

  • Easy to swallow
  • Film coated
  • Each Tablet contains Naproxen Sodium 220mg

Specifications

Product Type Mediproxen Tablets
Active Ingredients Naproxen Sodium 220 mg
For Use With (Application) Long-lasting relief from headaches, muscular aches, arthritis, fever
Quantity 50 x 1/Pk.
Recommended Age 12 Years and Older
Storage Requirements 68°F to 77°F (20°C to 25°C)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.