Learn More
Invitrogen™ NAPRT1 Monoclonal Antibody (CL0665)

Description
Immunogen sequence: PAFFEHLRAL DCSEVTVRAL PEGSLAFPGV PLLQVSGPLL VVQLLETPLL CLVSYASLVA TNAARLRLIA GP Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 96%. Binds to an epitope located within the peptide sequence PAFFEHLRALDCSEV as determined by overlapping synthetic peptides.
Specifications
Specifications
| Antigen | NAPRT1 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Clone | CL0665 |
| Concentration | 1.0 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | NAPRT |
| Gene Accession No. | Q6XQN6 |
| Gene Alias | 9130210N20Rik; FHA-HIT-interacting protein; FHIP; hypothetical protein MGC77870; Naprt; Naprt1; NAPRTase; Nicotinate phosphoribosyltransferase; nicotinate phosphoribosyltransferase domain containing 1; nicotinate phosphoribosyltransferase domain-containing protein 1; nicotinate phosphoribosyltransferase; LOW QUALITY PROTEIN: nicotinate phosphoribosyltransferase; nicotinate phosphoribosyltransferase-like protein; nicotinic acid phosphoribosyltransferase; PP3856; zgc:77870 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.