Learn More
Biogenex Laboratories Napsin A : IP64 : 6 mL
SDP
Supplier:  Biogenex Laboratories AM7015M
Encompass_Preferred
Napsin A has specific funtion in normal alveolar epithelim and is proposed to play a role in the protelytic processing of surfactant precursors. Napsin A is reported to be predominantly expressed in lamellar bodies of type II pneumocutes, secondary lysosymes of alveolar macrophages, respiratory epithelium of terminal and respiratory bronchioles, plasma cells within a subset of lymphocytes in normal lung, as well as in epithellial cells of renal tubiles in normal kidney and is weakly expressed in normal spleen
Catalog No. 50-197-8771
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.