Learn More
Abcam Narciclasine, Rho/LIM kinase/cofilin signaling pathway activator, 1MG
Supplier:  Abcam AB2181101MG
Encompass_Preferred
MW 307.25 Da, Purity >98%. Plant growth regulator originally isolated from Narcissus bulbs. Exhibits antiproliferative and pro-apoptotic effects in carcinoma cells and displays cytotoxic activity against a panel of 60 cancer cell lines (mean IC₅₀ = 47 nM). Activity decreases rate of cell division and increases mitosis duration in vitro. Also modulates the Rho/ROCK/LIM kinase/cofilin pathway; stimulates RhoA activation and induces actin polymerization.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-267-2827
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.