Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Naringenin 93.0+%, TCI America™
SDP

Catalog No. N007225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N007225G 25 g
N00725G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N007225G Supplier TCI America Supplier No. N007225G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 67604-48-2
Molecular Formula C15H12O5
Molecular Weight (g/mol) 272.256
MDL Number MFCD00006844
InChI Key FTVWIRXFELQLPI-UHFFFAOYSA-N
Synonym naringenin, 5,7-dihydroxy-2-4-hydroxyphenyl chroman-4-one, naringenine, 4',5,7-trihydroxyflavanone, +/--naringenin, naringetol, salipurpol, --naringenin, narigenin, salipurol
PubChem CID 932
ChEBI CHEBI:50202
IUPAC Name 5,7-dihydroxy-2-(4-hydroxyphenyl)-2,3-dihydrochromen-4-one
SMILES C1C(OC2=CC(=CC(=C2C1=O)O)O)C3=CC=C(C=C3)O

Specifications

Quantity 25 g
Formula Weight 272.26
Percent Purity ≥93.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Naringenin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.