Learn More
United Scientific™ UniStore™ Reagent Bottles, Narrow Mouth, HDPE

Shop All United Scientific Supplies, Inc Products


Description
UniStore™ Narrow Mouth, HDPE, reagent bottles are manufactured from USP Class VI resin that is FDA-21CFR compliant.
- Excellent chemical resistance to most acids, bases, and alcohols.
- Polypropylene caps are included.
- Manufactured from USP Class VI resin that is FDA-21CFR compliant for food contact and free of heavy metals.
- Leakproof closure with integrated seal ring.
- Precision molded for consistent wall thickness with smooth interior and exterior surfaces.
- Certificates of Analysis for leakproof test details are available upon request.
Specifications
Specifications
| Capacity (Metric) | 30 mL |
| Capacity (English) | 1 oz. |
| Color | Translucent |
| Material | HDPE |
| Packaging | Pack |
| Quantity | 72 Pk. |
| Sterility | Non-sterile |
| Product Line | UniStore |
| Closure Included | Yes |
| Graduated | No |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.