Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific™ UniStore™ Reagent Bottles, Narrow Mouth, HDPE, Bulk Packed
SDPSCI_ED

Catalog No. S43661 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Capacity (Metric):
60 mL
125 mL
250 mL
500 mL
1000 mL
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
S43661 1000 mL
S43654 60 mL
S43655 125 mL
S43656 250 mL
S43658 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. S43661 Supplier United Scientific™ Supplier No. 33405BULK
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Narrow Mouth HDPE Reagent Bottles are pilable, translucent and have fine contact clarity.

UniStore™ Narrow Mouth, HDPE, reagent bottles are manufactured from USP Class VI resin that is FDA-21CFR compliant.

  • Excellent chemical resistance to most acids, bases, and alcohols.
  • Polypropylene caps are included.
  • Manufactured from USP Class VI resin that is FDA-21CFR compliant for food contact and free of heavy metals.
  • Leakproof closure with integrated seal ring.
  • Precision molded for consistent wall thickness with smooth interior and exterior surfaces.
  • Certificates of Analysis for leakproof test details are available upon request.

Specifications

Capacity (Metric) 1000 mL
Color Translucent
Diameter (Metric) Inner Neck 21 mm
Material HDPE
No. per Case 50/Cs.
Quantity 50 Pk.
Sterility Non-sterile
Product Line UniStore
Closure Included Yes
Graduated No
Mouth Narrow
Height (Metric) 216 mm
Height (English) 8.5 in.
Closure Material Polypropylene
Closure Size 28 mm
Diameter (Metric) 21 mm
Through the Closure Dispensing No
Autoclavable Not Autoclavable
Capacity (English) 32 oz.
Product Type Reagent Bottle
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.