Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eisco Labs Narrow Mouth HDPE Translucent Reagent Bottles with Polypropylene Caps
SCI_ED

Catalog No. S000080 Shop All Eisco Products
Change view
Click to view available options
Capacity (Metric):
4 mL
8 mL
15 mL
30 mL
125 mL
500 mL
1 L
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
S000080 500 mL
S000075 4 mL
S000076 8 mL
S000077 15 mL
S000078 30 mL
S000079 125 mL
S000081 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. S000080 Supplier Eisco Labs Supplier No. CH5355PK12
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Narrow mouth translucent HDPE bottles with polypropylene caps offers leakproof, chemical-resistant storage that is ideal for safely handling reagents, samples, and solutions in lab or industrial environments.

  • Pack Of 12 HDPE Reagent Bottles: Includes narrow mouth translucent HDPE bottles with polypropylene caps, designed for secure storage and handling of liquids in laboratory or industrial settings.
  • Chemical Resistance: Made from HDPE, offering excellent resistance to a variety of chemicals, making these bottles ideal for diverse lab and storage applications.
  • Leak-Proof Tested: Rigorously tested to ensure leak protection during storage and transport. Features semi-buttress threads for a secure and reliable seal.
  • Temperature Range And Design: Operates from -100°C to +120°C. Translucent design allows visibility for liquid monitoring.
  • Versatile Applications: Suitable for storing reagents, samples, and solutions. Commonly used in laboratories, industrial facilities, and research settings for safe liquid containment.

Specifications

Capacity (Metric) 500 mL
Color Translucent
Diameter (Metric) Inner Neck 21.6 mm
Material HDPE
No. per Pack 12 Pk.
Sterility Non-sterile
Closure Included Yes
Graduated No
Mouth Narrow
Height (Metric) 170 mm
Diameter (Metric) 74 mm
Autoclavable Not Autoclavable
Capacity (English) 17 oz.
Product Type Reagent Bottle
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.