Learn More
Eisco Labs Narrow Mouth Polypropylene Reagent Bottles with Polypropylene Caps
Shop All Eisco Products

Description
- Narrow Mouth Bottles: Translucent polypropylene reagent bottles with polypropylene caps, designed for safe and efficient liquid storage in labs and industrial settings.
- Temperature And Chemical Resistance: Operates in a range of 0-135 °C and resists various chemicals, making it suitable for diverse lab and storage needs.
- Leak-Proof Design: Rigorously tested to prevent leaks during transport or storage. Features semi-buttress threads and secure polypropylene caps for tight closure.
- Autoclavable And Non-Sterile: Ideal for reusable lab workflows. Made from FDA-compliant and BSE/TSE-free materials.
- Lab And Industrial Applications: Suitable for research, sample storage, and reagent mixing. Translucent body provides visibility, and design ensures easy transport and handling.
Specifications
Specifications
| Capacity (Metric) | 500 mL |
| Color | Translucent |
| Material | Polypropylene |
| No. per Pack | 12 Pk. |
| Sterility | Non-sterile |
| Closure Included | Yes |
| Graduated | No |
| Mouth | Narrow |
| Height (Metric) | 170 mm |
| Diameter (Metric) | 74 mm |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.